Product References
A New Ligand-Based Method for Purifying Active Human Plasma-Derived Ficolin-3 Complexes Supports the Phenomenon of Crosstalk between Pattern-Recognition Molecules and Immunoglobulins.
PloS one
Man-Kupisinska A,Michalski M,Maciejewska A,Swierzko AS,Cedzynski M,Lugowski C,Lukasiewicz J
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Dot blot
Wed Jul 19 00:00:00 UTC 2017
Interaction of lectin pathway of complement-activating pattern recognition molecules with mycobacteria.
Clinical and experimental immunology
Bartlomiejczyk MA,Swierzko AS,Brzostek A,Dziadek J,Cedzynski M
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Sat Nov 01 00:00:00 UTC 2014
Interaction of lectin pathway of complement-activating pattern recognition molecules with mycobacteria.
Clinical and experimental immunology
Bartlomiejczyk MA,Swierzko AS,Brzostek A,Dziadek J,Cedzynski M
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Sat Nov 01 00:00:00 UTC 2014
Lectin-dependent enhancement of Ebola virus infection via soluble and transmembrane C-type lectin receptors.
PloS one
Brudner M,Karpel M,Lear C,Chen L,Yantosca LM,Scully C,Sarraju A,Sokolovska A,Zariffard MR,Eisen DP,Mungall BA,Kotton DN,Omari A,Huang IC,Farzan M,Takahashi K,Stuart L,Stahl GL,Ezekowitz AB,Spear GT,Olinger GG,Schmidt EV,Michelow IC
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in ELISA
Tue Oct 22 00:00:00 UTC 2013
The salivary scavenger and agglutinin binds MBL and regulates the lectin pathway of complement in solution and on surfaces.
Frontiers in immunology
Reichhardt MP,Loimaranta V,Thiel S,Finne J,Meri S,Jarva H
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in ELISA
Thu Aug 23 00:00:00 UTC 2012
A novel L-ficolin/mannose-binding lectin chimeric molecule with enhanced activity against Ebola virus.
The Journal of biological chemistry
Michelow IC,Dong M,Mungall BA,Yantosca LM,Lear C,Ji X,Karpel M,Rootes CL,Brudner M,Houen G,Eisen DP,Kinane TB,Takahashi K,Stahl GL,Olinger GG,Spear GT,Ezekowitz RA,Schmidt EV
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Fri Aug 06 00:00:00 UTC 2010
Characterization of the interaction between serum mannan-binding protein and nucleic acid ligands.
Journal of leukocyte biology
Nakamura N,Nonaka M,Ma BY,Matsumoto S,Kawasaki N,Asano S,Kawasaki T
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Tue Sep 01 00:00:00 UTC 2009
L-ficolin binding and lectin pathway activation by acetylated low-density lipoprotein.
Clinical and experimental immunology
Faro J,Chen Y,Jhaveri P,Oza P,Spear GT,Lint TF,Gewurz H
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Fri Feb 01 00:00:00 UTC 2008
Binding of serum mannan binding lectin to a cell integrity-defective Cryptococcus neoformans ccr4Delta mutant.
Infection and immunity
Panepinto JC,Komperda KW,Hacham M,Shin S,Liu X,Williamson PR
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Immunofluorescence
Mon Oct 01 00:00:00 UTC 2007
Alloiococcus otitidis is a ligand for collectins and Toll-like receptor 2, and its phagocytosis is enhanced by collectins.
European journal of immunology
Konishi M,Nishitani C,Mitsuzawa H,Shimizu T,Sano H,Harimaya A,Fujii N,Himi T,Kuroki Y
Published figure using MBL2 monoclonal antibody (Product # HYB 131-01-02) in Western Blot
Thu Jun 01 00:00:00 UTC 2006
Effect of stages of lactation on the concentration of a 90-kilodalton heat shock protein in bovine mammary tissue.
Journal of dairy science
Watanabe A,Miyamoto T,Katoh N,Takahashi Y
HYB 131-01-02 was used in western blot to investigate the differences of HSP90 expression in cow mammary tissue at various lactation stages
Wed Oct 01 00:00:00 UTC 1997